missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Pentylbenzolsulfonylchlorid, 97 %, Thermo Scientific™

Catalog Number p-7045482
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169677-20-7
C11H15ClO2S
246.749
MFCD00041509
WLBQFZZPRWQTGR-UHFFFAOYSA-N
4-tert-pentylbenzenesulfonyl chloride, 4-tert-amylbenzenesulphonyl chloride, 4-2-methylbutan-2-yl benzene-1-sulfonyl chloride, 4-tert-amylbenzenesulfonyl chloride, 4-tert-amyl-benzenesulphonyl chloride, 4-1,1-dimethylpropyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl benzenesulfonyl chloride, acmc-1bzun, 4-t-pentylbenzenesulfonyl chloride
22281976
4-(2-Methylbutan-2-yl)benzolsulfonylchlorid
CCC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169677-20-7
C11H15ClO2S
246.749
MFCD00041509
WLBQFZZPRWQTGR-UHFFFAOYSA-N
4-tert-pentylbenzenesulfonyl chloride, 4-tert-amylbenzenesulphonyl chloride, 4-2-methylbutan-2-yl benzene-1-sulfonyl chloride, 4-tert-amylbenzenesulfonyl chloride, 4-tert-amyl-benzenesulphonyl chloride, 4-1,1-dimethylpropyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl benzenesulfonyl chloride, acmc-1bzun, 4-t-pentylbenzenesulfonyl chloride
22281976
4-(2-Methylbutan-2-yl)benzolsulfonylchlorid
CCC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-tert-Pentylbenzenesulfonyl chloride
Schmelzpunkt 44°C to 49°C
Menge 1 g
UN-Nummer UN3261
Beilstein 2102813
Formelmasse 246.76
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'