Learn More
4-(Trifluoroacetyl)benzoic acid, 97+%
Product Code. 11398315
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398315 1 g 1g
11308325 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11398315 Supplier Thermo Scientific Alfa Aesar Supplier No. L10242.03
 Restricted Item
Verfügbar ab: 17-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 58808-59-6 | C9H5F3O3 | 218.131 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58808-59-6
Molecular Formula C9H5F3O3
Molecular Weight (g/mol) 218.131
MDL Number MFCD00052340
InChI Key WLTZCRCZDLLXQP-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
PubChem CID 143579
IUPAC Name 4-(2,2,2-trifluoroacetyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
Specifications
Melting Point 177°C to 179°C
Quantity 1 g
Beilstein 2728778
Sensitivity Hygroscopic
Formula Weight 218.13
Percent Purity ≥97%
Chemical Name or Material 4-(Trifluoroacetyl)benzoic acid