missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluormethoxy)benzylalkohol, 97 %, Thermo Scientific™

Catalog Number p-7024493
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1736-74-9
C8H7F3O2
192.137
MFCD00036029
ZLSOZAOCYJDPKX-UHFFFAOYSA-N
4-trifluoromethoxy benzyl alcohol, 4-trifluoromethoxy phenyl methanol, p-trifluoromethoxybenzyl alcohol, 4-trifluoromethoxy benzylalcohol, p-trifluoromethoxybenzylalchol, 4-trifluoromethoxybenzyl alcohol, 4-trifluoromethoxy-phenyl-methanol, benzenemethanol, 4-trifluoromethoxy, 4-trifluoromethoxy phenyl methan-1-ol
137184
[4-(trifluormethoxy)phenyl]methanol
C1=CC(=CC=C1CO)OC(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1736-74-9
C8H7F3O2
192.137
MFCD00036029
ZLSOZAOCYJDPKX-UHFFFAOYSA-N
4-trifluoromethoxy benzyl alcohol, 4-trifluoromethoxy phenyl methanol, p-trifluoromethoxybenzyl alcohol, 4-trifluoromethoxy benzylalcohol, p-trifluoromethoxybenzylalchol, 4-trifluoromethoxybenzyl alcohol, 4-trifluoromethoxy-phenyl-methanol, benzenemethanol, 4-trifluoromethoxy, 4-trifluoromethoxy phenyl methan-1-ol
137184
[4-(trifluormethoxy)phenyl]methanol
C1=CC(=CC=C1CO)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Trifluoromethoxy)benzyl alcohol
Dichte 1.326
Siedepunkt 100°C to 102°C (12 mmHg)
Flammpunkt 98°C (208°F)
Brechungsindex 1.449
Menge 1 g
Beilstein 1950379
Formelmasse 192.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'