missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluormethoxy)phenoxyessigsäure, 95 %, Thermo Scientific™

Catalog Number p-7033169
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72220-50-9
C9H7F3O4
236.15
MFCD00052352
QHSBEEUEIRDHCD-UHFFFAOYSA-N
2-4-trifluoromethoxy phenoxy acetic acid, 4-trifluoromethoxy phenoxyacetic acid, 4-trifluoromethoxy phenoxy acetic acid, 4-trifluoromethoxy-phenoxy-acetic acid, 2-4-trifluoromethyloxy phenoxy ethanoic acid, maybridge1_006105, acmc-20a5v1, 4-trifluoromethoxyphenoxyacetic acid
697705
2-[4-(trifluoromethoxy)phenoxy]acetic acid
OC(=O)COC1=CC=C(OC(F)(F)F)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72220-50-9
C9H7F3O4
236.15
MFCD00052352
QHSBEEUEIRDHCD-UHFFFAOYSA-N
2-4-trifluoromethoxy phenoxy acetic acid, 4-trifluoromethoxy phenoxyacetic acid, 4-trifluoromethoxy phenoxy acetic acid, 4-trifluoromethoxy-phenoxy-acetic acid, 2-4-trifluoromethyloxy phenoxy ethanoic acid, maybridge1_006105, acmc-20a5v1, 4-trifluoromethoxyphenoxyacetic acid
697705
2-[4-(trifluoromethoxy)phenoxy]acetic acid
OC(=O)COC1=CC=C(OC(F)(F)F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Trifluoromethoxy)phenoxyacetic acid
Menge 1 g
Formelmasse 236.15
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'