Learn More
Δ5(10),6,8(9)-D-(-)-Norgestrel, TRC
Specifications
Specifications
| Chemischer Name oder Material | 13-Ethyl-17-hydroxy-18,19-dinor-17Alpha-pregna-5,7,9-trien-20-yn-3-one |
| CAS | 155683-59-3 |
| Summenformel | C21 H24 O2 |
| Menge | 0.5 mg |
| Synonym | 18,19-Dinorpregna-5,7,9-trien-20-yn-3-one','13-ethyl-17-hydroxy-','(17α)-, Levonorgestrel Imp. W (EP), 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregna-5,7,9-trien-20-yn-3-one |
| InChI-Formel | InChI=1S/C21H24O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,5,7,19,23H,3,6,8-13H2,1H3/t19-,20-,21-/m0/s1 |
| SMILES | CC[C@]12CCc3c4CCC(=O)Cc4ccc3[C@@H]1CC[C@@]2(O)C#C |
| IUPAC-Name | (13S,14S,17R)-13-ethyl-17-ethinyl-17-Hydroxy-1,2,4,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-on |
| Molekulargewicht (g/mol) | 308.414 |
| Formelmasse | 308.178 |