missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Chlor-4-Nitrophenylcarbamoyl)-3-Fluorbenzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036252
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449132-61-9
C13H9BClFN2O5
338.482
MFCD20126189
JGVQEEYFVVEKHQ-UHFFFAOYSA-N
5-2-chloro-4-nitrophenylcarbamoyl-3-fluorobenzeneboronic acid, 3-2-chloro-4-nitrophenyl carbamoyl-5-fluorophenylboronic acid
73995848
[3-[(2-chlor-4-nitrophenyl)carbamoyl]-5-fluorphenyl]boronsäure
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])Cl)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449132-61-9
C13H9BClFN2O5
338.482
MFCD20126189
JGVQEEYFVVEKHQ-UHFFFAOYSA-N
5-2-chloro-4-nitrophenylcarbamoyl-3-fluorobenzeneboronic acid, 3-2-chloro-4-nitrophenyl carbamoyl-5-fluorophenylboronic acid
73995848
[3-[(2-chlor-4-nitrophenyl)carbamoyl]-5-fluorphenyl]boronsäure
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-(2-Chloro-4-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid
Menge 250 mg
Formelmasse 338.49
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'