missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Chlorpropyl)-3-methylisoxazol, TRC

Artikelnummer. 30355980
Change view
Click to view available options
Menge:
100 mg
Unit Size:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30355980 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355980 Supplier Toronto Research Chemicals Supplier No. TRCC379765100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 5-(3-Chloropropyl)-3-methylisoxazole
CAS 130800-76-9
Summenformel C7H10ClNO
Menge 100 mg
InChI-Formel InChI=1S/C7H10ClNO/c1-6-5-7(10-9-6)3-2-4-8/h5H,2-4H2,1H3
SMILES Cc1cc(CCCCl)on1
IUPAC-Name 5-(3-Chlorpropyl)-3-methyl-1,2-oxazol
Molekulargewicht (g/mol) 159.61
Formelmasse 159.0451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.