missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Pyridyl)-1H-tetrazol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7025320
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
14389-12-9
C6H5N5
147.141
MFCD00068115
JMILUUVWLRKJFB-UHFFFAOYSA-N
5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
482249
4-(2H-Tetrazol-5-yl)Pyridin
C1=CN=CC=C1C2=NNN=N2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
14389-12-9
C6H5N5
147.141
MFCD00068115
JMILUUVWLRKJFB-UHFFFAOYSA-N
5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
482249
4-(2H-Tetrazol-5-yl)Pyridin
C1=CN=CC=C1C2=NNN=N2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-(4-Pyridyl)-1H-tetrazole
Menge 1 g
Formelmasse 147.14
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'