missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(3-Chlorphenyl)-1H-Pyrazol-4-Carbonitril, 97 %, Thermo Scientific Chemicals

Catalog Number p-7029784
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
51516-68-8
C10H7ClN4
218.64
MFCD00128298
VRKRSWGTAMOYEV-UHFFFAOYSA-N
5-amino-1-3-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl-1h-pyrazole-, 5-amino-1-3-chloro-phenyl-1h-pyrazole-4-carbonit, 1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl, 1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl
734496
5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
51516-68-8
C10H7ClN4
218.64
MFCD00128298
VRKRSWGTAMOYEV-UHFFFAOYSA-N
5-amino-1-3-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl-1h-pyrazole-, 5-amino-1-3-chloro-phenyl-1h-pyrazole-4-carbonit, 1h-pyrazole-4-carbonitrile,5-amino-1-3-chlorophenyl, 1h-pyrazole-4-carbonitrile, 5-amino-1-3-chlorophenyl
734496
5-amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
NC1=C(C=NN1C1=CC=CC(Cl)=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Amino-1-(3-chlorophenyl)-1H-pyrazole-4-carbonitrile
Schmelzpunkt 183°C to 187°C
Menge 1 g
Formelmasse 218.65
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'