Learn More
5-Amino-2-methylbenzotrifluoride, TRC
Product Code. 30324087
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324087 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324087 Supplier Toronto Research Chemicals Supplier No. TRCA61524010G
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Amino-2-methylbenzotrifluoride
CAS 65934-74-9
Molecular Formula C8H8F3N
Quantity 10 g
Synonym (4-Methyl-3-trifluoromethylphenyl)amine, 3-Trifluoromethyl-4-methylaniline, 4-Amino-2-trifluoromethyltoluene, 4-Methyl-3-(trifluoromethyl)aniline, 4-Methyl-3-(trifluoromethyl)benzenamine
InChI Formula InChI=1S/C8H8F3N/c1-5-2-3-6(12)4-7(5)8(9,10)11/h2-4H,12H2,1H3
SMILES Cc1ccc(N)cc1C(F)(F)F
IUPAC Name 4-methyl-3-(trifluoromethyl)aniline
Molecular Weight (g/mol) 175.15
Formula Weight 175.0609