missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4 -Fluorophenyl)-1H-Pyrazol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7031676
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72411-52-0
C9H8FN3
177.18
MFCD01023677
QYEHDCXFXONDPV-UHFFFAOYSA-N
3-4-fluorophenyl-1h-pyrazol-5-amine, 5-4-fluorophenyl-1h-pyrazol-3-amine, 5-4-fluorophenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-fluorophenyl-1h-pyrazole, 5-amino-3-4-fluorophenyl pyrazole, 1h-pyrazol-3-amine,5-4-fluorophenyl, 5-4-fluorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-fluorophenyl-1h-pyrazole, 5-4-fluoro-phenyl-2h-pyrazol-3-ylamine, 3-4-fluorophenyl pyrazole-5-ylamine
2759138
5-(4-Fluorphenyl)-1H-Pyrazol-3-Amin
NC1=NNC(=C1)C1=CC=C(F)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
72411-52-0
C9H8FN3
177.18
MFCD01023677
QYEHDCXFXONDPV-UHFFFAOYSA-N
3-4-fluorophenyl-1h-pyrazol-5-amine, 5-4-fluorophenyl-1h-pyrazol-3-amine, 5-4-fluorophenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-fluorophenyl-1h-pyrazole, 5-amino-3-4-fluorophenyl pyrazole, 1h-pyrazol-3-amine,5-4-fluorophenyl, 5-4-fluorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-fluorophenyl-1h-pyrazole, 5-4-fluoro-phenyl-2h-pyrazol-3-ylamine, 3-4-fluorophenyl pyrazole-5-ylamine
2759138
5-(4-Fluorphenyl)-1H-Pyrazol-3-Amin
NC1=NNC(=C1)C1=CC=C(F)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Amino-3-(4-fluorophenyl)-1H-pyrazole
Schmelzpunkt 122°C to 125°C
Menge 1 g
Formelmasse 177.18
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'