missing translation for 'onlineSavingsMsg'
Learn More

5-Brom-2-(hydroxymethyl)-furo-[2,3-b]-pyridin, 98 %, Thermo Scientific™

Catalog Number 15332432
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1188990-02-4
C8H6BrNO2
228.05
MFCD13176658
SJASNTWQPIYYBY-UHFFFAOYSA-N
5-bromofuro 2,3-b pyridin-2-yl methanol, 5-bromo-2-hydroxymethyl furo 2,3-b pyridine
46318204
{5-bromofuro[2,3-b]pyridin-2-yl}methanol
OCC1=CC2=CC(Br)=CN=C2O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1188990-02-4
C8H6BrNO2
228.05
MFCD13176658
SJASNTWQPIYYBY-UHFFFAOYSA-N
5-bromofuro 2,3-b pyridin-2-yl methanol, 5-bromo-2-hydroxymethyl furo 2,3-b pyridine
46318204
{5-bromofuro[2,3-b]pyridin-2-yl}methanol
OCC1=CC2=CC(Br)=CN=C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Bromo-2-(hydroxymethyl)furo[2,3-b]pyridine
Schmelzpunkt 133°C to 135°C
Menge 5 g
Formelmasse 228.05
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'