Learn More
5-Brom-2-(Trifluoromethoxy)Toluol, 97 %, Thermo Scientific Chemicals
Catalog Number 15464797
Click to view available options
Menge:
250 mg
1 g
887268-26-0
C8H6BrF3O
255.03
MFCD04972695
PRSOCFCWLQJWBU-UHFFFAOYSA-N
4-bromo-2-methyl-1-trifluoromethoxy benzene, 5-bromo-2-trifluoromethoxytoluene, 5-bromo-2-trifluoromethoxy toluene, 3-methyl-4-trifluoromethoxy bromobenzene, 4-bromo-2-methyl trifluoromethoxy benzene, 4-bromo-2-methyl-trifluoromethoxy benzene, acmc-209qwv, 4-bromo-2-methyl-1-trifluoromethoxy-benzene, benzene,4-bromo-2-methyl-1-trifluoromethoxy
40427090
4-bromo-2-methyl-1-(trifluoromethoxy)benzene
CC1=C(OC(F)(F)F)C=CC(Br)=C1
887268-26-0
C8H6BrF3O
255.03
MFCD04972695
PRSOCFCWLQJWBU-UHFFFAOYSA-N
4-bromo-2-methyl-1-trifluoromethoxy benzene, 5-bromo-2-trifluoromethoxytoluene, 5-bromo-2-trifluoromethoxy toluene, 3-methyl-4-trifluoromethoxy bromobenzene, 4-bromo-2-methyl trifluoromethoxy benzene, 4-bromo-2-methyl-trifluoromethoxy benzene, acmc-209qwv, 4-bromo-2-methyl-1-trifluoromethoxy-benzene, benzene,4-bromo-2-methyl-1-trifluoromethoxy
40427090
4-bromo-2-methyl-1-(trifluoromethoxy)benzene
CC1=C(OC(F)(F)F)C=CC(Br)=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Bromo-2-(trifluoromethoxy)toluene
Menge 1 g
Formelmasse 255.04
Reinheit (%) 97%