Learn More
5-Brom-3-(3-oxycyclopentyl)-indol, 95 %, Thermo Scientific™
Catalog Number p-7055232
Click to view available options
Menge:
250 mg
1 g
676170-02-8
C13H12BrNO
278.149
OGXFMBGFSFOQGH-UHFFFAOYSA-N
3-5-bromo-1h-indol-3-yl cyclopentanone, 3-5-bromo-1h-indole-3-yl cyclopentanone, 3-5-bromo-1h-indol-3-yl cyclopentan-1-one
22139469
3-(5-Brom-1H-Indol-3-yl)Cyclopentan-1-on
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)Br
676170-02-8
C13H12BrNO
278.149
OGXFMBGFSFOQGH-UHFFFAOYSA-N
3-5-bromo-1h-indol-3-yl cyclopentanone, 3-5-bromo-1h-indole-3-yl cyclopentanone, 3-5-bromo-1h-indol-3-yl cyclopentan-1-one
22139469
3-(5-Brom-1H-Indol-3-yl)Cyclopentan-1-on
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)Br
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Bromo-3-(3-oxycyclopentyl)indole
Menge 250 mg
Formelmasse 278.14
Reinheit (%) 95%
Physikalische Form Fest