missing translation for 'onlineSavingsMsg'
Learn More

5-Brom-N-methylpyridin-2-carboxamid, 96 %, Thermo Scientific Chemicals

Catalog Number 15401088
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
845305-87-5
C7H7BrN2O
215.05
MFCD07357421
OHYCXPXWHPWYFX-UHFFFAOYSA-N
5-bromo-n-methylpicolinamide, n-methyl 5-bromopicolinamide, 2-pyridinecarboxamide, 5-bromo-n-methyl, n-methyl5-bromopicolinamide, acmc-209pv5, 5-bromo-n-methylpicolimide, n-methyl-5-bromopicolinamide, 5-bromo-n-methyl-2-pyridinecarboxamide
22831620
5-Brom-N-Methylpyridin-2-Carboxamid
CNC(=O)C1=NC=C(C=C1)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
845305-87-5
C7H7BrN2O
215.05
MFCD07357421
OHYCXPXWHPWYFX-UHFFFAOYSA-N
5-bromo-n-methylpicolinamide, n-methyl 5-bromopicolinamide, 2-pyridinecarboxamide, 5-bromo-n-methyl, n-methyl5-bromopicolinamide, acmc-209pv5, 5-bromo-n-methylpicolimide, n-methyl-5-bromopicolinamide, 5-bromo-n-methyl-2-pyridinecarboxamide
22831620
5-Brom-N-Methylpyridin-2-Carboxamid
CNC(=O)C1=NC=C(C=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Bromo-N-methylpyridine-2-carboxamide
Schmelzpunkt 76°C to 77°C
Menge 1 g
Formelmasse 215.05
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'