missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoyl-2-Fluorbenzolboronsäure-Pinacolester, 96 %, Thermo Scientific™

Catalog Number p-7039812
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1412905-42-0
C13H17BFNO3
265.09
MFCD18730404
RBEAFMIZDXOHAT-UHFFFAOYSA-N
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester
99738340
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1412905-42-0
C13H17BFNO3
265.09
MFCD18730404
RBEAFMIZDXOHAT-UHFFFAOYSA-N
4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 5-carbamoyl-2-fluorobenzeneboronic acid pinacol ester
99738340
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(=C1)C(N)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester
Menge 250 mg
Formelmasse 265.09
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'