missing translation for 'onlineSavingsMsg'
Learn More

5'-Fluor-2'-(Trifluormethyl)acetophenon, 97 %, Thermo Scientific™

Catalog Number p-7031190
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
243863-39-0
C9H6F4O
206.14
MFCD00236274
USFYRRQTKUSMNK-UHFFFAOYSA-N
5'-fluoro-2'-trifluoromethyl acetophenone, 1-5-fluoro-2-trifluoromethyl phenyl ethanone, 5'-fluoro-2'-trifluoromethyl acetophenon, 1-acetyl-5-fluoro-2-trifluoromethyl benzene
2774766
1-[5 -Fluor-2-(Trifluormethyl)phenyl]Ethanon
CC(=O)C1=C(C=CC(=C1)F)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
243863-39-0
C9H6F4O
206.14
MFCD00236274
USFYRRQTKUSMNK-UHFFFAOYSA-N
5'-fluoro-2'-trifluoromethyl acetophenone, 1-5-fluoro-2-trifluoromethyl phenyl ethanone, 5'-fluoro-2'-trifluoromethyl acetophenon, 1-acetyl-5-fluoro-2-trifluoromethyl benzene
2774766
1-[5 -Fluor-2-(Trifluormethyl)phenyl]Ethanon
CC(=O)C1=C(C=CC(=C1)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5'-Fluoro-2'-(trifluoromethyl)acetophenone
Brechungsindex 1.4462
Menge 1 g
Formelmasse 206.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'