missing translation for 'onlineSavingsMsg'
Learn More

5'-Fluoroindirubinoxime, Tocris Bioscience™

FLT3 inhibitor; displays antiproliferative activity

Marke:  Tocris (Bio-Techne) 403310

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413994

  • 190.08 € / 10 Milligramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Inhibitor of FMS-like receptor tyrosine kinase-3 (FLT3) (IC50 = 15 nM). Displays selectivity for FLT3 against 6 other kinases including EGFR. Displays antiproliferative activity against the MV4;11 cell line (expressing constitutively active FLT3) and a number of cancer cell lines, including SNU-638 (stomach carcinoma) and HT-1080 (fibrosarcoma).
Spezifikation

Chemische Identifikatoren

861214-33-7
295.273
5'-fluoroindirubinoxime, 5'-fluoro-1h,1'h-2,3' biindolylidene-3,2'dione 3-oxime, 5/'-fluoroindirubinoxime, d0t9fe, a05-a4b1-ii, 3z-5-fluoro-3-3e-3-hydroxyiminoindolin-2-ylidene indolin-2-one, 5-fluoro-3-3-hydroxyimino-1,3-dihydro-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
5-fluoro-3-[3-(hydroxyamino)-1H-indol-2-yl]indol-2-one
C16H10FN3O2
LBUXOYSDWHGJNP-UHFFFAOYSA-N
11580208
C1=CC=C2C(=C1)C(=C(N2)C3=C4C=C(C=CC4=NC3=O)F)NO

Spezifikation

5'-Fluoroindirubinoxime
861214-33-7
5'-fluoroindirubinoxime, 5'-fluoro-1h,1'h-2,3' biindolylidene-3,2'dione 3-oxime, 5/'-fluoroindirubinoxime, d0t9fe, a05-a4b1-ii, 3z-5-fluoro-3-3e-3-hydroxyiminoindolin-2-ylidene indolin-2-one, 5-fluoro-3-3-hydroxyimino-1,3-dihydro-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
C1=CC=C2C(=C1)C(=C(N2)C3=C4C=C(C=CC4=NC3=O)F)NO
295.273
295.27
10mg
C16H10FN3O2
LBUXOYSDWHGJNP-UHFFFAOYSA-N
5-fluoro-3-[3-(hydroxyamino)-1H-indol-2-yl]indol-2-one
11580208
>99%
Videos