missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorsalicylaldehyd, +98 %, Thermo Scientific Chemicals

Catalog Number p-7059041
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
347-54-6
C7H5FO2
140.113
MFCD01090997
FDUBQNUDZOGOFE-UHFFFAOYSA-N
5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
2737328
5-Fluor-2-Hydroxybenzaldehyd
C1=CC(=C(C=C1F)C=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
347-54-6
C7H5FO2
140.113
MFCD01090997
FDUBQNUDZOGOFE-UHFFFAOYSA-N
5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
2737328
5-Fluor-2-Hydroxybenzaldehyd
C1=CC(=C(C=C1F)C=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Fluorosalicylaldehyde
Schmelzpunkt 82°C to 85°C
Menge 1 g
Empfindlichkeit Air Sensitive
Formelmasse 140.12
Reinheit (%) ≥98%
Physikalische Form Pulver oder Flocken

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'