Learn More
5-Methyl-3-heptanone, TRC
Product Code. 30478741
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478741 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30478741 Supplier Toronto Research Chemicals Supplier No. TRCM312070250ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methyl-3-Heptanone
CAS 541-85-5
Molecular Formula C8 H16 O
Quantity 250 mL
Synonym 3-Heptanone','5-methyl-, 5-Methyl-3-heptanone, (±)-5-Methyl-3-heptanone, 2-Methylbutyl ethyl ketone, 3-Methyl-5-heptanone, Ethyl 2-methylbutyl ketone, NSC 100734
InChI Formula InChI=1S/C8H16O/c1-4-7(3)6-8(9)5-2/h7H,4-6H2,1-3H3
SMILES CCC(C)CC(=O)CC
IUPAC Name 5-methylheptan-3-one
Molecular Weight (g/mol) 128.21
Formula Weight 128.1201