Learn More
5-Methyl Cytosine Hydrochloride, TRC
Product Code. 30468500
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468500 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30468500 Supplier Toronto Research Chemicals Supplier No. TRCM2947005G
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methylcytosine Hydrochloride
CAS 58366-64-6
Molecular Formula C5 H7 N3 O . Cl H
Quantity 5 g
Synonym 2(1H)-Pyrimidinone, 6-amino-5-methyl-, hydrochloride (1:1), 2(1H)-Pyrimidinone, 4-amino-5-methyl-, monohydrochloride (9CI), Cytosine, 5-methyl-, hydrochloride (6CI), 5-Methylcytosine hydrochloride, 6-Amino-5-methyl-2(1H)-pyrimidinone hydrochloride, 4-Amino-2-hydroxy-5-methylpyrimidine hydrochloride, 4-Amino-5-methyl-2(1H)-pyrimidinone hydrochloride, 5-Methyl cytosine hydrochloride
InChI Formula InChI=1S/C5H7N3O.ClH/c1-3-2-7-5(9)8-4(3)6;/h2H,1H3,(H3,6,7,8,9);1H
SMILES Cl.CC1=CNC(=O)N=C1N
IUPAC Name 4-amino-5-methyl-1H-pyrimidin-2-one;hydrochloride
Molecular Weight (g/mol) 161.59
Formula Weight 161.0356