missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylpyrazin-2-carbonsäure, 97 %, Thermo Scientific™
Click to view available options
Menge:
5 g
10 g
25 g
Spezifikation
Chemischer Name oder Material | 5-Methylpyrazine-2-carboxylic acid |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Menge | 5 g |
Formelmasse | 138.13 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'