missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylthiophen-2-boronsäurepinakolester, 95 %, Thermo Scientific™

Catalog Number 15464736
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
459409-74-6
C16H19BO2S
286.196
MFCD08063141
BOTJOCNWKJVWRR-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-5-phenylthiophen-2-yl-1,3,2-dioxaborolane, 5-phenylthiophene-2-boronic acid pinacol ester, 2-phenylthiophene-5-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-phenylthiophene-2-yl-1,3,2-dioxaborolane, 5-phenylthiophene-2-boronic acid pinacol este, 5-phenylthiophen-2-yl boronic acid pinacol ester, 2-5-phenyl-2-thienyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-phenyl-2-thienyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-phenyl-thiophen-2-yl-1,3,2-dioxaborolane
45786345
4,4,5,5-Tetramethyl-2-(5-Phenylthiophen-2-yl)-1,3,2-Dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C3=CC=CC=C3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
459409-74-6
C16H19BO2S
286.196
MFCD08063141
BOTJOCNWKJVWRR-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-5-phenylthiophen-2-yl-1,3,2-dioxaborolane, 5-phenylthiophene-2-boronic acid pinacol ester, 2-phenylthiophene-5-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-5-phenylthiophene-2-yl-1,3,2-dioxaborolane, 5-phenylthiophene-2-boronic acid pinacol este, 5-phenylthiophen-2-yl boronic acid pinacol ester, 2-5-phenyl-2-thienyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-phenyl-2-thienyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-phenyl-thiophen-2-yl-1,3,2-dioxaborolane
45786345
4,4,5,5-Tetramethyl-2-(5-Phenylthiophen-2-yl)-1,3,2-Dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 5-Phenylthiophene-2-boronic acid pinacol ester
Menge 1 g
Formelmasse 286.2
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'