missing translation for 'onlineSavingsMsg'
Learn More

6-(1-Pyrrolidinyl)pyridin-3-Carboxaldehyd, 98 %, Thermo Scientific Chemicals

Catalog Number p-7033114
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261715-39-3
C10H12N2O
176.219
MFCD06200881
FFFAXGTZBUMKTI-UHFFFAOYSA-N
6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
2794775
6-Pyrrolidin-1-ylpyridin-3-Carbaldehyd
C1CCN(C1)C2=NC=C(C=C2)C=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261715-39-3
C10H12N2O
176.219
MFCD06200881
FFFAXGTZBUMKTI-UHFFFAOYSA-N
6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
2794775
6-Pyrrolidin-1-ylpyridin-3-Carbaldehyd
C1CCN(C1)C2=NC=C(C=C2)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-(1-Pyrrolidinyl)pyridine-3-carboxaldehyde
Menge 1 g
Empfindlichkeit Air Sensitive
Formelmasse 176.22
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'