missing translation for 'onlineSavingsMsg'
Learn More

6,10,11-Triethylcarbonat Daunomycinon, TRC

Product Code. 30450906
Change view
Click to view available options
Menge:
1 mg
Unit Size:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30450906 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450906 Supplier Toronto Research Chemicals Supplier No. TRCT7763801MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 6,10,11-Triethylcarbonate Daunomycinone
CAS 67665-61-6
Summenformel C30H30O14
Menge 1 mg
Synonym (1S-cis)-3-Acetyl-1,2,3,4,6,11-hexahydro-3-hydroxy-10-methoxy-6,11-dioxo-1,5,12-naphthacenetriyl Carbonic Acid Triethyl Ester
InChI-Formel InChI=1S/C30H30O14/c1-6-39-27(34)42-18-13-30(37,14(4)31)12-16-20(18)26(44-29(36)41-8-3)22-21(25(16)43-28(35)40-7-2)23(32)15-10-9-11-17(38-5)19(15)24(22)33/h9-11,18,37H,6-8,12-13H2,1-5H3/t18-,30-/m0/s1
SMILES CCOC(=O)O[C@H]1C[C@@](O)(Cc2c(OC(=O)OCC)c3C(=O)c4cccc(OC)c4C(=O)c3c(OC(=O)OCC)c12)C(=O)C
IUPAC-Name [(1S,3S)-3-Acetyl-1,12-bis(ethoxycarbonyloxy)-3-hydroxy-10-methoxy-6,11-dioxo-2,4-dihydro-1H-tetracen-5-yl]ethylcarbonat
Molekulargewicht (g/mol) 614.55
Formelmasse 614.1636
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.