Learn More
6-(4-Bromphenyl)-4,5-dihydro-3(2H)-pyridazinon, 98 %, Thermo Scientific™
Catalog Number p-7033468
Click to view available options
Menge:
1 g
5 g
36725-37-8
C10H9BrN2O
253.099
MFCD00203490
CDDNLMGOLYMTSF-UHFFFAOYSA-N
6-4-bromophenyl-4,5-dihydropyridazin-3 2h-one, 6-4-bromophenyl-4,5-dihydro-2h-pyridazin-3-one, 6-4-bromophenyl-2,3,4,5-tetrahydropyridazin-3-one, 6-4-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 6-4-bromophenyl-2,4,5-trihydropyridazin-3-one, 3-4-bromophenyl-4,5-dihydro-1h-pyridazin-6-one, 6-4-bromophenyl-4 5-dihydro-2h-pyrida, 6-p-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 3 2h-pyridazinone,6-4-bromophenyl-4,5-dihydro, 6-4-bromo-phenyl-4,5-dihydro-2h-pyridazin-3-one
689108
3-(4-Bromphenyl)-4,5-Dihydro-1H-Pyridazin-6-on
C1CC(=O)NN=C1C2=CC=C(C=C2)Br
36725-37-8
C10H9BrN2O
253.099
MFCD00203490
CDDNLMGOLYMTSF-UHFFFAOYSA-N
6-4-bromophenyl-4,5-dihydropyridazin-3 2h-one, 6-4-bromophenyl-4,5-dihydro-2h-pyridazin-3-one, 6-4-bromophenyl-2,3,4,5-tetrahydropyridazin-3-one, 6-4-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 6-4-bromophenyl-2,4,5-trihydropyridazin-3-one, 3-4-bromophenyl-4,5-dihydro-1h-pyridazin-6-one, 6-4-bromophenyl-4 5-dihydro-2h-pyrida, 6-p-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 3 2h-pyridazinone,6-4-bromophenyl-4,5-dihydro, 6-4-bromo-phenyl-4,5-dihydro-2h-pyridazin-3-one
689108
3-(4-Bromphenyl)-4,5-Dihydro-1H-Pyridazin-6-on
C1CC(=O)NN=C1C2=CC=C(C=C2)Br
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone
Schmelzpunkt 167°C to 171°C
Menge 1 g
Formelmasse 253.1
Reinheit (%) 98%