Learn More
6-Brom-1,8-naphthyridin-2(1H)-on, 98 %, Thermo Scientific Chemicals
Catalog Number p-7033037
Click to view available options
Menge:
250 mg
1 g
72754-05-3
C8H5BrN2O
225.05
MFCD07781151
BCEWGGGNOQNYKK-UHFFFAOYSA-N
12520144
6-Brom-1H-1,8-Naphthyridin-2-on
BrC1=CN=C2NC(=O)C=CC2=C1
72754-05-3
C8H5BrN2O
225.05
MFCD07781151
BCEWGGGNOQNYKK-UHFFFAOYSA-N
12520144
6-Brom-1H-1,8-Naphthyridin-2-on
BrC1=CN=C2NC(=O)C=CC2=C1
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
6-Bromo-1,8-Naphthyridin-2(1H)-one wird als organisches chemisches Synthese-Zwischenprodukt verwendet.
Löslichkeit
Unlöslich in Wasser.
Hinweise
Stabil unter empfohlenen Lagerbedingungen. Nicht kompatibel mit Oxidationsmitteln.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Bromo-1,8-naphthyridin-2(1H)-one
Schmelzpunkt >300°C
Menge 250 mg
Synonym 6-bromo-1,8-naphthyridin-2-ol, 6-bromo-1,8-naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2-ol, 6-bromo-2-hydroxy-1,8-naphthyridine, 1,8-naphthyridin-2 1h-one, 6-bromo, acmc-1bihh, ksc496s1t, 6-bromopyridino 2,3-b pyridin-2-ol, 6-bromo-1,8naphthyridin-2 1h-one
Löslichkeitsinformationen Insoluble in water.
Formelmasse 225.05
Reinheit (%) 98%