missing translation for 'onlineSavingsMsg'
Learn More

6-Brom-4-methylchinolin, 96 %, Thermo Scientific Chemicals

Catalog Number p-7031991
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41037-28-9
C10H8BrN
222.085
MFCD09258893
XBZQSJLYRMZRSZ-UHFFFAOYSA-N
quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
12475362
6-Brom-4-Methylchinolin
CC1=C2C=C(C=CC2=NC=C1)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41037-28-9
C10H8BrN
222.085
MFCD09258893
XBZQSJLYRMZRSZ-UHFFFAOYSA-N
quinoline, 6-bromo-4-methyl, 6-bromo-4-methyl-quinoline, 6-bromo lepidine, 4-methyl-6-bromoquinoline, quinoline,6-bromo-4-methyl, 6-bromanyl-4-methyl-quinoline
12475362
6-Brom-4-Methylchinolin
CC1=C2C=C(C=CC2=NC=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Bromo-4-methylquinoline
Schmelzpunkt 94°C to 95°C
Menge 250 mg
Empfindlichkeit Air and light sensitive
Formelmasse 222.08
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'