missing translation for 'onlineSavingsMsg'
Learn More

6-Brompyridin-2-carboxamid, 97 %, Thermo Scientific Chemicals

Catalog Number 15464447
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25194-52-9
C6H5BrN2O
201.02
MFCD00234154
HWRUOHMDYQEWOD-UHFFFAOYSA-N
21961408
6-Brompyridin-2-Carboxamid
NC(=O)C1=NC(Br)=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
25194-52-9
C6H5BrN2O
201.02
MFCD00234154
HWRUOHMDYQEWOD-UHFFFAOYSA-N
21961408
6-Brompyridin-2-Carboxamid
NC(=O)C1=NC(Br)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Bromopyridine-2-carboxamide
Menge 1 g
Synonym 6-bromopicolinamide, 6-bromopyridine-2-carboxylic acid amide, 6-bromo-2-pyridinecarboxamide, 2-pyridinecarboxamide, 6-bromo, 6-bromo-pyridine-2-carboxylic acid amide, 6-bromo-2-carbamoylpyridine, 2-pyridinecarboxamide,6-bromo, acmc-209gi0, tpc-i169, 6-bromanylpyridine-2-carboxamide
Formelmasse 201.02
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'