missing translation for 'onlineSavingsMsg'
Learn More

6-Chlor-2-Fluor-3-Methoxyphenylessigsäure, 97 %, Thermo Scientific™

Catalog Number p-7031583
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1017777-83-1
C9H8ClFO3
218.608
MFCD09832273
SZVJYTRCVDHYHX-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methoxyphenylacetic acid, 2-6-chloro-2-fluoro-3-methoxy-phenyl acetic acid, 6-chloro-2-fluoro-3-methoxyphenyl acetic acid, 2-6-chloro-2-fluoro-3-methoxyphenyl acetic acid
46737509
2-(6-Chlor-2-Fluor-3-methoxyphenyl)essigsäure
COC1=C(C(=C(C=C1)Cl)CC(=O)O)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1017777-83-1
C9H8ClFO3
218.608
MFCD09832273
SZVJYTRCVDHYHX-UHFFFAOYSA-N
6-chloro-2-fluoro-3-methoxyphenylacetic acid, 2-6-chloro-2-fluoro-3-methoxy-phenyl acetic acid, 6-chloro-2-fluoro-3-methoxyphenyl acetic acid, 2-6-chloro-2-fluoro-3-methoxyphenyl acetic acid
46737509
2-(6-Chlor-2-Fluor-3-methoxyphenyl)essigsäure
COC1=C(C(=C(C=C1)Cl)CC(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Chloro-2-fluoro-3-methoxyphenylacetic acid
Schmelzpunkt 134°C to 137°C
Menge 250 mg
Formelmasse 218.61
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'