missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorooxindol, 98%, Thermo Scientific Chemicals

Catalog Number p-7024642
Click to view available options
Menge:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56341-37-8
C8H6ClNO
167.592
MFCD00209962
CENVPIZOTHULGJ-UHFFFAOYSA-N
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
736344
6-Chlor-1,3-dihydro-2-indolon
C1C2=C(C=C(C=C2)Cl)NC1=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
56341-37-8
C8H6ClNO
167.592
MFCD00209962
CENVPIZOTHULGJ-UHFFFAOYSA-N
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one
736344
6-Chlor-1,3-dihydro-2-indolon
C1C2=C(C=C(C=C2)Cl)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Chlorooxindole
Schmelzpunkt 195°C to 199°C
Menge 5 g
Formelmasse 167.6
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'