missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxypyridin-3-boronsäurepinakolester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7034026
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1054483-78-1
C11H16BNO3
221.06
MFCD06795671
WAUWXCUPDOXYKS-UHFFFAOYSA-N
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2 1h-one, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb145, pyridin-2-ol-5-boronic acid pinacol ester
17750253
5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)-1H-Pyridin-2-on
CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1054483-78-1
C11H16BNO3
221.06
MFCD06795671
WAUWXCUPDOXYKS-UHFFFAOYSA-N
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2 1h-one, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb145, pyridin-2-ol-5-boronic acid pinacol ester
17750253
5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)-1H-Pyridin-2-on
CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Hydroxypyridine-3-boronic acid pinacol ester
Menge 250 mg
Formelmasse 221.06
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'