missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-propionitril, 97 %, Thermo Scientific™

Catalog Number p-7029705
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
351003-19-5
C12H12N2O2
216.24
MFCD03427105
SZEGFLHUNYLAOJ-UHFFFAOYSA-N
3-6-methyl-2h-1,4-benzoxazin-3 4h-one-4-yl-propionitrile, 3-6-methyl-3-oxo-2h-1,4-benzoxazin-4-yl propanenitrile, 3-6-methyl-2h-1 4-benzoxazin-3 4h-one, 3-6-methyl-3-oxo-1,4-benzoxazin-4-yl propanenitrile, 4h-1,4-benzoxazine-4-propanenitrile,2,3-dihydro-6-methyl-3-oxo, 6-methyl-3-oxo-2,3-dihydro-4h-1,4-benzoxazine-4-propionitrile, 3-2,3-dihydro-6-methyl-3-oxobenzo b 1,4 oxazin-4-yl propanenitrile, 3-6-methyl-3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl propanenitrile, 3-6-methyl-3-oxo-2h-benzo b 1,4 oxazin-4 3h-yl propanenitrile
4524694
3-(6-Methyl-3-Oxo-1,4-Benzoxazin-4-yl)Propannitril
CC1=CC2=C(C=C1)OCC(=O)N2CCC#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
351003-19-5
C12H12N2O2
216.24
MFCD03427105
SZEGFLHUNYLAOJ-UHFFFAOYSA-N
3-6-methyl-2h-1,4-benzoxazin-3 4h-one-4-yl-propionitrile, 3-6-methyl-3-oxo-2h-1,4-benzoxazin-4-yl propanenitrile, 3-6-methyl-2h-1 4-benzoxazin-3 4h-one, 3-6-methyl-3-oxo-1,4-benzoxazin-4-yl propanenitrile, 4h-1,4-benzoxazine-4-propanenitrile,2,3-dihydro-6-methyl-3-oxo, 6-methyl-3-oxo-2,3-dihydro-4h-1,4-benzoxazine-4-propionitrile, 3-2,3-dihydro-6-methyl-3-oxobenzo b 1,4 oxazin-4-yl propanenitrile, 3-6-methyl-3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl propanenitrile, 3-6-methyl-3-oxo-2h-benzo b 1,4 oxazin-4 3h-yl propanenitrile
4524694
3-(6-Methyl-3-Oxo-1,4-Benzoxazin-4-yl)Propannitril
CC1=CC2=C(C=C1)OCC(=O)N2CCC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 6-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-propionitrile
Schmelzpunkt 128°C to 130°C
Menge 1 g
Formelmasse 216.24
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'