Learn More
600 mOsm/kg Osmolality Standard, Reagecon
BIOPHARMA
Product Code. 16339221 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
12 x 5 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16339221 12 x 5 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16339221 Supplier Reagecon Supplier No. REOSM600
  • ONLINE EXCLUSIVE PRICE
    180.00 €
    140.32 € / 60 Milliliter
    Sparen 39.68 €
    22% Rabatt
 Restricted Item
Lager vorrätig
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Used to calibrate, control or validate osmometers
  • Manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines
  • Tested by ISO 17025 accredited test method
  • ISO 17025 accreditation for calibration of laboratory balances
  • Prepared gravimetrically on weight/weight basis
Specifications
Packaging Amber glass ampoule
Physical Form Liquid
Technique Calibration, Control or Validate Osmometer
For Use With (Equipment) Osmometer
Type Osmolality Standard
Quantity 12 x 5 mL