missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyflavon, 99 %, Thermo Scientific™

Catalog Number p-7027819
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
22395-22-8
C16H12O3
252.269
MFCD00017462
QKNDCRMJDZLFEG-UHFFFAOYSA-N
7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one
466268
7-Methoxy-2-phenylchromen-4-on
COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
22395-22-8
C16H12O3
252.269
MFCD00017462
QKNDCRMJDZLFEG-UHFFFAOYSA-N
7-methoxyflavone, 7-methoxy-2-phenyl-4h-chromen-4-one, flavone, 7-methoxy, unii-xk561ap5ur, 7-methoxy-2-phenyl-chromen-4-one, xk561ap5ur, 7-methoxy-2-phenylchromone, 7-methoxy flavone, 7-methoxy-2-phenyl-4h-benzopyran-4-one
466268
7-Methoxy-2-phenylchromen-4-on
COC1=CC2=C(C=C1)C(=O)C=C(O2)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 7-Methoxyflavone
Schmelzpunkt 108°C to 112°C
Menge 1 g
Formelmasse 252.27
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'