missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1,2,3,4-tetrahydroisochinolin Hydrochlorid, ≥ 97 %, Thermo Scientific Chemicals

Catalog Number p-7034693
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
99365-69-2
C9H11ClN2O2
214.649
MFCD07371494
KGIXLJRTYZOUCW-UHFFFAOYSA-N
7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-7-nitroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 1,2,3,4-tetrahydro-7-nitro-, monohydrochloride, ksc683k9f, 7-nitro-1,2,3,4-tetrahydroisoquinoline monohydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
13521670
7-Nitro-1,2,3,4-Tetrahydroisochinolin;Hydrochlorid
C1CNCC2=C1C=CC(=C2)[N+](=O)[O-].Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
99365-69-2
C9H11ClN2O2
214.649
MFCD07371494
KGIXLJRTYZOUCW-UHFFFAOYSA-N
7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-7-nitroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 1,2,3,4-tetrahydro-7-nitro-, monohydrochloride, ksc683k9f, 7-nitro-1,2,3,4-tetrahydroisoquinoline monohydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
13521670
7-Nitro-1,2,3,4-Tetrahydroisochinolin;Hydrochlorid
C1CNCC2=C1C=CC(=C2)[N+](=O)[O-].Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Menge 250 mg
Formelmasse 214.65
Reinheit (%) ≥97%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'