missing translation for 'onlineSavingsMsg'
Learn More

8-Chlor-6-(2-fluorphenyl)-1-methyl-4H-imidazo[1,5-a][1,4]benzodiazepin-5-oxid (Midazolam-5-oxid), TRC

Artikelnummer. 30351051
Change view
Click to view available options
Menge:
1 mg
Unit Size:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30351051 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351051 Supplier Toronto Research Chemicals Supplier No. TRCC3664251MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Midazolam 5-Oxide
CAS 59468-83-6
Hinweise zur Reinheitsqualität HPLC
Summenformel C18 H13 Cl F N3 O
Menge 1 mg
Synonym 8-Chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5-a][1,4]benzodiazepine 5-Oxide, Midazolam 5-Oxide, Midazolam Imp. D (EP)
InChI-Formel InChI=1S/C18H13ClFN3O/c1-11-21-9-13-10-22(24)18(14-4-2-3-5-16(14)20)15-8-12(19)6-7-17(15)23(11)13/h2-9H,10H2,1H3
SMILES Cc1ncc2C[N+](=C(c3ccccc3F)c4cc(Cl)ccc4n12)[O-]
IUPAC-Name 8-Chlor-6-(2-Fluorphenyl)-1-methyl-5-oxido-4H-imidazo[1,5-a][1,4]benzodiazepin-5-ium
Molekulargewicht (g/mol) 341.77
Formelmasse 341.0731
Reinheit (%) >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.