missing translation for 'onlineSavingsMsg'
Learn More

8-Chlor-6-(trifluormethyl)-imidazo-[1,2-a]-pyridin, 97 %, Thermo Scientific™

Catalog Number p-7032826
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
178488-36-3
C8H4ClF3N2
220.579
MFCD06496224
BWSHPRLSCABXRL-UHFFFAOYSA-N
8-chloro-6-trifluoromethyl imidazo 1,2-a pyridine, chlorotrifluoromethylimidazopyridine, 8-chloro-6-trifluoromethyl-4-hydroimidazo 1,2-a pyridine
5260659
8-Chlor-6-(Trifluormethyl)Imidazo[1,2-a]Pyridin
C1=CN2C=C(C=C(C2=N1)Cl)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
178488-36-3
C8H4ClF3N2
220.579
MFCD06496224
BWSHPRLSCABXRL-UHFFFAOYSA-N
8-chloro-6-trifluoromethyl imidazo 1,2-a pyridine, chlorotrifluoromethylimidazopyridine, 8-chloro-6-trifluoromethyl-4-hydroimidazo 1,2-a pyridine
5260659
8-Chlor-6-(Trifluormethyl)Imidazo[1,2-a]Pyridin
C1=CN2C=C(C=C(C2=N1)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Schmelzpunkt 100°C to 103°C
Menge 1 g
Formelmasse 220.58
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'