missing translation for 'onlineSavingsMsg'
Learn More

8-(Trifluormethyl)-imidazo-[1,2-a]-pyridin-2-carbonsäure Hydrat, 95 %, Thermo Scientific™

Catalog Number p-7032902
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1018828-72-2
C9H5F3N2O2
230.15
MFCD09995161
QRXSLIVFFDRFEB-UHFFFAOYSA-N
8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid
59443885
8-(Trifluormethyl)Imidazo[1,2-a]Pyridin-2-Carbonsäure
OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1018828-72-2
C9H5F3N2O2
230.15
MFCD09995161
QRXSLIVFFDRFEB-UHFFFAOYSA-N
8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid
59443885
8-(Trifluormethyl)Imidazo[1,2-a]Pyridin-2-Carbonsäure
OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Schmelzpunkt ∽242°C (decomposition)
Menge 250 mg
Formelmasse 230.15 (Anhydrous)
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'