missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 9-cis Retinoic Acid

Product Code. 17140062
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 17140062

Brand: STEMCELL Technologies 72384

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy

Retinoid pathway activator; Activates retinoic acid receptor (RAR) and retinoid X receptor (RXR)

  • 9-cis-Retinoic acid activates all isoforms of the retinoic acid receptor (RAR) (Ki = 0.5-27 nM) as well as retinoid X receptor (RXR) isoforms (Ki = 3.8-12 nM) (Umemiya et al.; Wong et al.) The ability of RAR to heterodimerize with RXR, and the latter to heterodimerize with other partners allows 9-cis-retinoic acid to evoke a wide range of effects (Dawson et al.; Kane)

Specifications

CAS 5300-03-8
Synonym 9-​cis-​retinoic acid; Aliretinoin; NSC 659772; Panretin
Quantity 25 mg
Molecular Formula C20H28O2
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.