Learn More
9-Mesityl-10-methylacridinium Tetrafluoroborate, TRC
Specifications
Specifications
| Chemical Name or Material | 9-Mesityl-10-methylacridinium Tetrafluoroborate |
| CAS | 1442433-71-7 |
| Molecular Formula | C23H22BF4N |
| Quantity | 500 mg |
| Synonym | 10-Methyl-9-(2,4,6-trimethylphenyl)-acridinium Tetrafluoroborate(1-) (1:1) |
| InChI Formula | InChI=1S/C23H22N.BF4/c1-15-13-16(2)22(17(3)14-15)23-18-9-5-7-11-20(18)24(4)21-12-8-6-10-19(21)23;2-1(3,4)5/h5-14H,1-4H3;/q+1;-1 |
| SMILES | C[N+]1=C(C=CC=C2)C2=C(C3=C(C)C=C(C)C=C3C)C4=CC=CC=C41.FB(F)F.[F-] |
| IUPAC Name | 10-methyl-9-(2,4,6-trimethylphenyl)acridin-10-ium;tetrafluoroborate |
| Molecular Weight (g/mol) | 399.23 |
| Formula Weight | 399.1781 |