Learn More
TPP™ Gewebekulturbehandelte 96-Well Mikrotiterplatten
Beschreibung
Color: Clear; Sterility; Sterile; Lid With Lid
- Air-venting system in lid guarantees controlled gas exchange with low evaporation; vents in plate base prevent condensation and provide consistent air-flow between stacked plates
- Sloped edge allows placement of the lid in one position only; yellow marking area on lid and plate ensure correct lid orientation
- Ridged grip area ensures a secure grasp and prevents accidental lifting of lid
- Easy to see black alpha-numerical labelling between the wells for easy orientation during microscopy
- Growth area on spherical zone only; absolutely flat growth surface
- Plate sizes (6, 12, 24, 48 and 96-well) stack securely together
- Manufactured meeting the standards ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004
- White base colour is embossed firmly on the test plate eliminating loss of identification through detachment
- 2 well geometries available: F-base and U-base
Spezifikation
Spezifikation
| Barcode | Alphanumerisch |
| Format | Standard |
| Deckel | Mit Deckel |
| Kulturfläche | 0.322 cm2 |
| Array | 8 x 12 |
| Verpackung | 6 Stück/Packung; 108 Stück/Karton |
| Farbe | Clear |
| Material | Polystyrol |
| Anzahl Wells | 96 |
| Sterilität | Steril |
| Mehr anzeigen |