Learn More
Tocris Bioscience™ 968
Allosteric inhibitor of glutaminase
N.V.
Beschreibung
Allosteric inhibitor of glutaminase. Blocks Rho-GTPase-dependent transformation of fibroblasts. Suppresses breast cancer cell growth and invasive activity in vitro. Also inhibits P-493 B lymphoma cell growth and reduces tumor size of P-493 cell xenografts in mice.Spezifikation
968 | |
968 | |
475.42 |
For Research Use Only