missing translation for 'onlineSavingsMsg'
Learn More

A 1120, Tocris Bioscience™

High affinity retinol-binding protein 4 (RBP4) ligand

Marke:  Tocris (Bio-Techne) 379310

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413614

  • 231.01 € / 10 Milligramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

High affinity retinol-binding protein 4 (RBP4) ligand (Ki = 8.3 nM); non-retinoid. Selective against a range of different cellular targets. As efficacious as fenretinide (Cat. No. 1396) in the reduction of serum RBP4 and retinol. Displaces transthyretin (TTR) from RBP4-TTR complexes.
Spezifikation

Chemische Identifikatoren

1152782-19-8
392.378
compound 33 pmid: 24835984, 2-4-2-trifluoromethyl phenyl piperidine-1-carboxamido benzoic acid, 2-4-2-trifluoromethyl phenyl piperidine-1-carbonylamino benzoic acid, a hplc , powder, 2-4-2-trifluoromethyl phenyl piperidine-1-carboxamido benzoic acid, 2-4-2-trifluoromethyl phenyl piperidine-1-carbonyl amino benzoic acid, 2-4-2-trifluoromethyl phenyl piperidin-1-yl carbonyl amino benzoic acid
2-[[4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonyl]amino]benzoic acid
C20H19F3N2O3
MEAQCLPMSVEOQF-UHFFFAOYSA-N
25138295
C1CN(CCC1C2=CC=CC=C2C(F)(F)F)C(=O)NC3=CC=CC=C3C(=O)O

Spezifikation

A 1120
1152782-19-8
compound 33 pmid: 24835984, 2-4-2-trifluoromethyl phenyl piperidine-1-carboxamido benzoic acid, 2-4-2-trifluoromethyl phenyl piperidine-1-carbonylamino benzoic acid, a hplc , powder, 2-4-2-trifluoromethyl phenyl piperidine-1-carboxamido benzoic acid, 2-4-2-trifluoromethyl phenyl piperidine-1-carbonyl amino benzoic acid, 2-4-2-trifluoromethyl phenyl piperidin-1-yl carbonyl amino benzoic acid
C1CN(CCC1C2=CC=CC=C2C(F)(F)F)C(=O)NC3=CC=CC=C3C(=O)O
392.378
392.37
10mg
C20H19F3N2O3
MEAQCLPMSVEOQF-UHFFFAOYSA-N
2-[[4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonyl]amino]benzoic acid
25138295
>99%
Videos