Learn More
Kimberly-Clark™ Professional A25+ Kleenguard™ SMS-Overall
Provides general protection against water-based or chemical splashes, liquids or dust particles. Kimberly Clark™ KLEENGUARD™ Hooded A25+ Splash Protection Coveralls are perfect for general manufacturing and maintenance areas.
Marke:  Kimberly-Clark™ Professional 89810
Weitere Details : Netto-Gewicht : 2.50000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418218
  • 185.00 € / 25 Stück
Nicht verfügbar
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Ideal for working in general manufacturing and maintenance
  • Offers complete freedom of movement with comfortable body design
  • Protects against powders, larger fibers, radioactive dust and chemical splashes
  • Silicone-free and anti-static
Spezifikation
Spezifikation
Elastic
2X-Large
2.5Kg
Men's
Elastic
128 to 136cm
White
Elastic
Coveralls
Full-length Zips
Hooded
Videos