missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
AARSD1 Polyclonal specifically detects AARSD1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Spezifikation
Spezifikation
| Antigen | AARSD1 |
| Anwendungen | Western Blot, ELISA |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:32000 |
| Zusammensetzung | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gen-Zugriffsnummer | NP_001129514.2 |
| Gen-Alias | alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 |
| Gensymbole | AARSD1 |
| Wirtsspezies | Goat |
| Mehr anzeigen |
For Research Use Only
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?