missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14996-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function.Spezifikation
ABCB6 | |
Polyclonal | |
Unconjugated | |
ABCB6 | |
ABC, ABC14, ABCB6, EST45597, Mt ABC transporter 3, MTABC3, P glycoprotein related protein, PRP, umat | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10058, 140669, 74104 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70595, Q9DC29, Q9NP58 | |
Abcb6 | |
ABCB6 Fusion Protein Ag6982 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |