Learn More
ABCF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556456
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CVWLEEELKT FKRILVLVSH SQDFLNGVCT NIIHMHNKKL KYYTGNYDQY VKTRLELEEN QMKRFHWEQD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ABCF2 Polyclonal antibody specifically detects ABCF2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ABCF2 | |
Polyclonal | |
Unconjugated | |
Abcf2 | |
0710005O05Rik; ABC28; Abcf2; ABC-type transport protein; ATP binding cassette subfamily F member 2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, subfamily F (GCN20), member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; DKFZp586K1823; Drr3; E430001O06; EST133090; HUSSY18; HUSSY-18; Iron-inhibited ABC transporter 2; M-ABC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10061, 27407, 311959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99LE6, Q9UG63 | |
Abcf2 | |
Recombinant protein corresponding to Human ABCF2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only