Learn More
ABCG4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559139
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NPMLFRAVQN GLCAMAEKKS SPEKNEVPAP CPPCPPEVDP IESHTFATST LT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.
ABCG4 Polyclonal antibody specifically detects ABCG4 in Human samples. It is validated for Immunohistochemistry.Spezifikation
ABCG4 | |
Polyclonal | |
Unconjugated | |
Abcg4 | |
6430517O04Rik; ABCG4; ATP binding cassette subfamily G member 4; ATP-binding cassette sub-family G member 4; ATP-binding cassette, subfamily G (WHITE), member 4; ATP-binding cassette, sub-family G (WHITE), member 4; putative ABC transporter; WHITE2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H172 | |
Abcg4 | |
Recombinant protein corresponding to Human ABCG4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only