Learn More
ABHD12B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551760
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LGTGVATNAA KVLEEKGCPV DAIVLEAPFT NMWVASINYP LLKIYRNIPG FLRTLMDALR KDKIVFPNDE NVKFLSSPLL ILHGEDDRTV PLEYGKKLYE IARNAYRNKE RVKMVIFPPG FQHNLLCKSP TLLITVRDFL SKQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 44%.
ABHD12B Polyclonal antibody specifically detects ABHD12B in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ABHD12B | |
Polyclonal | |
Unconjugated | |
ABHD12B | |
ABHD12B; abhydrolase domain containing 12B; abhydrolase domain-containing protein 12B; Alpha/beta hydrolase domain-containing protein 12B; Bem46l3; c14_5314; C14orf29; Gm804; Protein ABHD12B; serine protease BEM46-like 3; Variant 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145447 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5M8 | |
ABHD12B | |
Recombinant protein corresponding to Human ABHD12B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only